15-Crown-5
Synonym(s):
1,4,7,10,13-Pentaoxacyclopentadecane
Empirical Formula (Hill Notation):
C10H20O5
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing