Skip to Content
Merck
All Photos(1)

Key Documents

1040683

USP

Apigenin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4′,5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
262620
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

apigenin

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H

InChI key

KZNIFHPLKGYRTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flavonoids belong to polyphenolic class of compounds. They play crucial role in biological activities which include tumor growth inhibition and chemoprevention. Apigenin is a plant flavonoid mostly used to inhibit ornithine decarboxylase activity and skin tumor.

Application

Apigenin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Powdered Soy Isoflavones Extract
  • Soy Isoflavones Capsules
  • Soy Isoflavones Tablets

Biochem/physiol Actions

A plant flavonoid that has been found to inhibit cell proliferation by arresting the cell cycle at the G2/M phase. Inhibition of growth through cell cycle arrest and induction of apoptosis appear to be related to induction of p53. Inhibits PMA-mediated tumor promotion by inhibiting protein kinase C and the resulting suppression of oncogene expression. It has also been reported to inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service