Skip to Content
Merck
All Photos(1)

Key Documents

58067

Nylon 66 filter membrane filter discs .45 μm pore size

pore size 0.45 μm, diam. 47 mm, pkg of 50 ea

Synonym(s):

MEMBRANES NYLON 66 .45UM PK/50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
eCl@ss:
32110203

product name

Nylon 66 Filter Membranes, pore size 0.45 μm, diam. 47 mm, pkg of 50 ea

material

Nylon 66 filter membrane

packaging

pkg of 50 ea

diam.

47 mm

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

Nylon 66 Filter Membranes protect the instrument and columns by removing particles and gases from solvents and other mobile phase components. They are compatible with all solvents commonly used in HPLC. For use with sink aspirator.

Application

Nylon 66 filter membranes have been used in filtering the reagents such as acetonitrile and methanol used as mobile phase in the determination of aflatoxins in rice samples using high-performance liquid chromatography with fluorescence detection (HPLC-FL).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Post-harvest operations and aflatoxin levels in rice (Oryza sativa).
Prietto L, et al.
Crop Protection, 78, 172-177 (2015)
Parshuram J Sonawane et al.
PloS one, 6(1), e16661-e16661 (2011-02-10)
3-Hydroxy-3-methylglutaryl-coenzyme A [HMG-CoA] reductase gene (Hmgcr) is a susceptibility gene for essential hypertension. Sequencing of the Hmgcr locus in genetically hypertensive BPH (blood pressure high), genetically hypotensive BPL (blood pressure low) and genetically normotensive BPN (blood pressure normal) mice yielded

Articles

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service