Skip to Content
Merck
All Photos(3)

Key Documents

WH0006570M1

Sigma-Aldrich

Monoclonal Anti-SLC18A1 antibody produced in mouse

clone 3A2-E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CGAT, Anti-VAT1, Anti-VMAT1, Anti-solute carrier family 18 (vesicular monoamine), member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A2-E10, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC18A1(6570)

Related Categories

General description

The vesicular monoamine transporter acts to accumulate cytosolic monoamines into vesicles, using the proton gradient maintained across the vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine (Peter et al., 1993 [PubMed 7905859]). See also SLC18A2 (MIM 193001).[supplied by OMIM

Immunogen

SLC18A1 (AAH06317.1, 1 a.a. ~ 493 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLRPILDAPQRLLKEGRASRQLVLVVVFVALLLDNMLFTVVVPIVPTFLYDMEFKEVNSSLHLGHAGSSPHALASPAFSTIFSFFNNNTVAVEESVPSGIAWMNDTASTIPPPATEAISAHKNNCLQGTGFLEEETTRVGVLFASKAVMQLLVNPFVGPLTNRIGYHIPMFAGFVIMFLSTVMFAFSGTYTLLFVARTLQGIGSSFSSVAGLGMLASVYTDDHERGRAMGTALGGLALGLLVGAPFGSVMYEFVGKSAPFLILAFLALLDGALQLCILQPSKVSPESAKGTPLFMLLKDPYILVAAGLAFLPASVSYLIGTNLFGVLANKMGRWLCSLIGMLVVGTSLLCVPLAHNIFGLIGPNAGLGLAIGMVDSSMMPIMGHLVDLRHTSVYGSVYAIADVAFCMGFAIGPSTGGAIVKAIGFPWLMVITGVINIVYAPLCYYLRSPPAKEEKLAILSQDCPMETRMYATQKPTKEFPLGEDSDEEPDHEE

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service