Skip to Content
Merck
All Photos(3)

Documents

T5910

Sigma-Aldrich

2,3,5-Triiodobenzoic acid

BioReagent, suitable for plant cell culture, ≥97% (HPLC)

Synonym(s):

TIBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I3C6H2CO2H
CAS Number:
Molecular Weight:
499.81
Beilstein:
1955088
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

≥97% (HPLC)

technique(s)

cell culture | plant: suitable

mp

220-222 °C (lit.)

application(s)

agriculture

storage temp.

−20°C

SMILES string

OC(=O)c1cc(I)cc(I)c1I

InChI

1S/C7H3I3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)

InChI key

ZMZGFLUUZLELNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,5-Triiodobenzoic acid has been used as a supplement of MS media to study the auxin distribution and transport during embryogenesis and seed germination of Arabidopsis.

Biochem/physiol Actions

2,3,5-Triiodobenzoic acid (TIBA) is an auxin transport inhibitor and is used to study the role of auxin flow during plant growth and development.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Auxin regulates first leaf development and promotes the formation of protocorm trichomes and rhizome-like structures in developing seedlings of Spathoglottis plicata (Orchidaceae)
Novak SD and Whitehouse GA
AoB PLANTS, 5(4), 169-169 (2013)
Acropetal auxin transport inhibition is involved in indeterminate but not determinate nodule formation
Ng JLP, et al.
Frontiers in Plant Science, 9(4), 169-169 (2018)
Pankaj Dhonukshe et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(11), 4489-4494 (2008-03-14)
Many aspects of plant development, including patterning and tropisms, are largely dependent on the asymmetric distribution of the plant signaling molecule auxin. Auxin transport inhibitors (ATIs), which interfere with directional auxin transport, have been essential tools in formulating this concept.
Hardik Kundariya et al.
Nature communications, 11(1), 5343-5343 (2020-10-24)
Plants transmit signals long distances, as evidenced in grafting experiments that create distinct rootstock-scion junctions. Noncoding small RNA is a signaling molecule that is graft transmissible, participating in RNA-directed DNA methylation; but the meiotic transmissibility of graft-mediated epigenetic changes remains
Auxin distribution and transport during embryogenesis and seed germination of Arabidopsis
Ni DA, et al.
Cell Research, 11(4), 273-273 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service