Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700609

Sigma-Aldrich

Monoclonal Anti-TRAT1-PE antibody produced in mouse

clone TRIM-04, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-T cell receptor-interacting molecule, Anti-TRIM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TRIM-04, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

0.1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TRAT1(50852)

General description

The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells.

Immunogen

Recombinant intracellular domain (aa 29-186) of human TRIM

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:50. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service