Skip to Content
Merck
All Photos(1)

Key Documents

MAK480

Sigma-Aldrich

Ethanol Assay Kit

sufficient for 500 colorimetric tests

Synonym(s):

Ethanol Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

food and beverages

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

shipped in

ambient

storage temp.

2-8°C

General description

Ethanol is a psychoactive component of many commonly consumed drinks where it acts as a central nervous system depressant. After ingestion, ethanol is absorbed into the bloodstream via the stomach and small intestine. Ethanol is largely metabolized by the liver but is also secreted in urine or through respiration. The monitoring of ethanol levels is also important in fermentation processes. Studies have shown that heavy alcohol consumption may lead to various forms of liver diseases and to increased mortality rates.

Features and Benefits

  • The linear detection range of the kit is 0.04 to 2.0%.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the quantitative determination of ethanol in samples such as alcoholic beverages.

Principle

The Ethanol Assay Kit is based on an improved dichromate method, in which dichromate is reduced by ethanol to a bluish chromic (Cr3+) product. The intensity of color, measured at 580 nm, is a direct measure of the alcohol concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples and exhibits high sensitivity.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service