Skip to Content
Merck
All Photos(1)

Key Documents

HPA027887

Sigma-Aldrich

Anti-NAV1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DKFZp781D0314, Anti-FLJ12560, Anti-FLJ14203, Anti-KIAA1151, Anti-MGC14961, Anti-POMFIL3, Anti-neuron navigator 1, Anti-steerin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

AKSFVKPPSLANLDKVNSNSLDLPSSSDTTHASKVPDLHATSSASGGPLPSCFTPSPAPILNINSASFSQGLELMSGFSVPKETRMYPKLSGLHRSMESLQMPMSLPSAFPSSTPVPTPPAPPAAPTEEETEELTWSGSP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAV1(89796)

General description

Neuron navigator 1 (NAV1) is expressed in the brain during the developmental stages and in the adult heart. The gene encoding this protein is localized on human chromosome 1q32.1.

Immunogen

neuron navigator 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Neuron navigator 1 (NAV1) interacts with TRIO (a rho guanine nucleotide exchange factor) and is involved in neurite outgrowth. This interaction increases the affinity of TRIO for Ras-related C3 botulinum toxin substrate 1 (Rac1) and Ras homolog family member G (RhoG) which are involved in the process.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72802

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuron navigator: a human gene family with homology to unc-53, a cell guidance gene from Caenorhabditis elegans.
Maes T, et.al
Genomics, 80(1), 21-30 (2002)
Dynamic microtubules catalyze formation of navigator-TRIO complexes to regulate neurite extension.
van Haren J, et.al
Current Biology, 24(15), 1778-1785 (2014)
Jeffrey van Haren et al.
Current biology : CB, 24(15), 1778-1785 (2014-07-30)
Neurite extension is regulated by multiple signaling cascades that ultimately converge on the actin and microtubule networks [1]. Rho GTPases, molecular switches that oscillate between an inactive, GDP-bound state and an active, GTP-bound state, play a pivotal role in controlling
Tamara Maes et al.
Genomics, 80(1), 21-30 (2002-06-25)
We have cloned the gene neuron navigator-1 (NAV1), a human homolog of unc-53, a gene involved in axon guidance in Caenorhabditis elegans. Duplications during evolution gave rise to three human homologs located on chromosomes 1q32.1, 11p15.1, and 12q21.1. NAV1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service