Skip to Content
Merck
All Photos(7)

Key Documents

HPA001527

Sigma-Aldrich

Anti-TF antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-β-1-Metal-binding globulin antibody produced in rabbit, Anti-Serotransferrin precursor antibody produced in rabbit, Anti-Siderophilin antibody produced in rabbit, Anti-Transferrin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DGPSVACVKKASYLDCIRAIAANEADAVTLDAGLVYDAYLAPNNLKPVVAEFYGSKEDPQTFYYAVAVVKKDSGFQMNQLRGKKSCHTGLGRSAGWNIPIGLLYCDLPEPRKPLEKA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TF(7018)

Looking for similar products? Visit Product Comparison Guide

General description

TF (transferrin) is a 80kDa monomeric blood plasma glycoprotein with iron bindinding property. It is braodly divded into two regions with two iron binding sites. Both the N-terminal and C-terminal domains consist of two sub domains (N1 and N2, C1 and C2) connected by a hinge region.

Immunogen

Serotransferrin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TF antibody produced in rabbit is suitable for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-TF antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

TF (transferrin) gene encodes a glycoprotein that binds to two Fe3+ ions in association with the binding of an anion, usually bicarbonate. It functions in transporting iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. Most of the bound iron is utilized for the synthesis of hemoglobin by developing erythroid cells. It may also facilitate the removal of certain organic matter and allergens from serum by binding to granulocyte/pollen. Defects in this gene are associated with atransferrinemia, characterized by abnormal synthesis of transferrin leading to iron overload and microcytic hypochromic anemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84506

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Martin et al.
Clinical chemistry, 33(3), 405-407 (1987-03-01)
An understanding of Al3+-induced diseases requires identification of the blood carrier of Al3+ to the tissues where Al3+ exerts a toxic action. Quantitative studies demonstrate that the protein transferrin (iron-free) is the strongest Al3+ binder in blood plasma. Under plasma
Wenqiang Ma et al.
The British journal of nutrition, 117(7), 923-929 (2017-05-24)
To investigate the effects of maternal dietary protein restriction on offspring Fe metabolism, twenty-four second-parity Landrace×Yorkshire sows were randomly allocated to standard-protein (SP) and low-protein (LP) groups. The SP sows were fed diets containing 15 and 18 % crude protein
Pablo F Garcia et al.
Biochimica et biophysica acta, 1840(9), 2695-2701 (2014-06-28)
Transferrin is an iron-binding blood plasma glycoprotein that controls the level of free iron in biological fluids. This protein has been deeply studied in the past few years because of its potential use as a strategy of drug targeting to
P Ponka et al.
Seminars in hematology, 35(1), 35-54 (1998-02-14)
Iron represents a paradox for living systems by being essential for a wide variety of metabolic processes (oxygen transport, electron transport, DNA synthesis, etc) but also having the potential to cause deleterious effects. Because of Iron's virtual insolubility and potential
Huifang Li et al.
Nutrients, 9(6) (2017-06-21)
Exposure to stress is known to cause hepatic iron dysregulation, but the relationship between prolonged stress and liver iron metabolism is not yet fully understood. Thirty 13-week-old female Sprague-Dawley rats were randomly divided into two groups, as follows: the control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service