Skip to Content
Merck
All Photos(1)

Key Documents

EP0030089871

Eppendorf® Combitips advanced® pipette tips, Forensic DNA Grade

blue, pack of 100 ea (individually wrapped)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

material

blue

sterility

non-sterile

packaging

pack of 100 ea (individually wrapped)

manufacturer/tradename

Eppendorf® 0030089871

maximum volume

5 mL

color

blue

Looking for similar products? Visit Product Comparison Guide

Application

Eppendorf® Combitips® Advanced pipette tips, Forensic DNA Grade are suitable for:

  • DNAextraction
  • sample processing
  • polymerase chain reaction (PCR) setup

Features and Benefits

  • Eppendorf® Forensic DNA Grade consumables exemplify the highest quality and performance.
  • The test parameters and sensitivity levels correspond to the requirements of the ISO18385 standard and are subject to strict controls.
  • Tested for the absence of DNase, RNase, and PCR inhibitor

Other Notes

Browse all pipette tips

Legal Information

Combitips is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service