Skip to Content
Merck
All Photos(1)

Key Documents

EHU036851

Sigma-Aldrich

MISSION® esiRNA

targeting human NTS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCTCCTGGAGTCTGTGCTCAGATTCAGAAGAGGAAATGAAAGCATTAGAAGCAGATTTCTTGACCAATATGCATACATCAAAGATTAGTAAAGCACATGTTCCCTCTTGGAAGATGACTCTGCTAAATGTTTGCAGTCTTGTAAATAATTTGAACAGCCCAGCTGAGGAAACAGGAGAAGTTCATGAAGAGGAGCTTGTTGCAAGAAGGAAACTTCCTACTGCTTTAGATGGCTTTAGCTTGGAAGCAATGTTGACAATATACCAGCTCCACAAAATCTGTCACAGCAGGGCTTTTCAACACTGGGAGTTAATCCAGGAAGATATTCTTGATACTGGAAATGACAAAAATGGAAAGGAAGAAGTCATAAAGAGAAAAATTCCTTATATTCTGAAACGGCAGCTGTATGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhang et al.
Molecular therapy. Nucleic acids, 14, 262-273 (2019-01-18)
Development of the receptive endometrium (RE) from the pre-receptive endometrium (PE) is essential for embryo implantation, but its molecular mechanisms have not been fully understood. In this study, lncRNA-miRNA-mRNA and circRNA-miRNA-mRNA networks were constructed to explore the functions of potential competing endogenous
Brett Verstak et al.
Journal of leukocyte biology, 96(3), 427-436 (2014-05-09)
TLRs act as sentinels in professional immune cells to detect and initiate the innate immune response to pathogen challenge. TLR4 is a widely expressed TLR, responsible for initiating potent immune responses to LPS. TRAM acts to bridge TLR4 with TRIF
Xiao-Yu Shi et al.
Asian Pacific journal of tropical medicine, 7(10), 787-791 (2014-08-19)
To explore the effect of Fibulin-5 expression on cell proliferation and invasion in human gastric cancer patients. Fibulin-5 expression was detected in 56 samples of surgically resected gastric cancer and paired noncancerous tissues using qRT-PCR and immunoblotting. Fibulin-5 was knocked
J K Liu et al.
Cell death and differentiation, 21(8), 1325-1339 (2014-05-17)
Glioblastoma is the most common primary intrinsic brain tumor and remains incurable despite maximal therapy. Glioblastomas display cellular hierarchies with self-renewing glioma-initiating cells (GICs) at the apex. To discover new GIC targets, we used in vivo delivery of phage display
Yuqi Huang et al.
International journal of molecular sciences, 15(10), 18148-18161 (2014-10-11)
Targeting protein for Xenopus kinesin-like protein 2 (TPX2), a microtubule-associated protein, impacts spindle assembly in human cells. Several studies have demonstrated that TPX2 is overexpressed in different types of human cancers and promotes tumor growth and metastasis. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service