Skip to Content
Merck
All Photos(1)

Key Documents

COLO 668

87061209, human lung (brain metastasis), Round & Refractile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COLO 668, from human lung(brain metastasis), 87061209

biological source

human lung (brain metastasis)

growth mode

Suspension

karyotype

2n = 46

morphology

Round & Refractile

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung oat cell carcinoma

Cell Line Description

Derived from a brain metastasis of a 47 year old female with oat cell carcinoma of the lung.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 10,11
D16S539: 11,12
D5S818: 11
D7S820: 8.2,10
THO1: 9
TPOX: 8
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service