Skip to Content
Merck
All Photos(1)

Key Documents

52033

Sigma-Aldrich

UV-VIS detectable IEF Marker solution

pI 9.6, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Synonym(s):

IEF Marker UV-vis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105336
NACRES:
NA.26

sterility

0.45 μm filtered

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as IEF marker for HPCE and IEF gels.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service