Skip to Content
Merck
All Photos(1)

Key Documents

SMB01327

Sigma-Aldrich

5-Methoxytryptophol

Synonym(s):

2-(5-Methoxy-1H-indol-3-yl)ethanol, 3-(2-Hydroxyethyl)-5-methoxyindole, 5-Methoxy-1H-indole-3-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

Assay

95% (HPLC)

Quality Level

form

liquid

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]cc(CCO)c2c1

InChI

1S/C11H13NO2/c1-14-9-2-3-11-10(6-9)8(4-5-13)7-12-11/h2-3,6-7,12-13H,4-5H2,1H3

InChI key

QLWKTGDEPLRFAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methoxytryptophol is a serotonin metabolite. It is synthesized by the pineal gland and together with melatonin is involved with diurnal rhythms regulation.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service