Skip to Content
Merck
All Photos(1)

Key Documents

PHR1464

Supelco

Dapsone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Aminophenyl sulfone, 4,4′-Diaminodiphenyl sulfone, 4,4′-Sulfonyldianiline, Bis(4-aminophenyl) sulfone, DDS, Dapsone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NC6H4)2SO2
CAS Number:
Molecular Weight:
248.30
Beilstein:
788055
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 102
traceable to Ph. Eur. D0100000
traceable to USP 1164008

API family

dapsone

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

175-177 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)c2ccc(N)cc2

InChI

1S/C12H12N2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H,13-14H2

InChI key

MQJKPEGWNLWLTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dapsone is an antibacterial drug, belongs to a class of compounds known as sulfones. It acts by inhibiting the synthesis of dihydrofolic acid through competition with para-aminobenzoate for the active site of dihydropteroate synthetase, thereby inhibiting the growth of microorganisms that are dependent on endogenous folic acid synthesis. It is used in primary treatment of dermatitis herpetiformis and in combination with other drugs to treat leprosy.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dapsone may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3012 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 1B - STOT RE 2 - STOT SE 2

Target Organs

Blood, Blood,spleen,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dapsone and sulfones in dermatology: overview and update
Zhu YI and Stiller MJ
Journal of the American Academy of Dermatology, 45(3), 420-434 (2001)
Validated TLC and HPLC stability-indicating methods for the quantitative determination of dapsone
Salama N, et al.
JPC-Journal of Planar Chromatography-Modern TLC, 25(1), 65-71 (2012)
An ecofriendly and stability-indicating RP-HPLC method for determination of dapsone: Application to pharmaceutical analysis
Beheshti ML, et al.
Chiang Mai Journal of Science, 43(3), 620-629 (2016)
Chunjuan Yang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 960, 59-66 (2014-05-03)
Corynoline and acetycorynoline are active compounds of Corydalis bungeana Turcz. with various pharmacological effects such as sedation, anti-leptospira and liver injury protection effects. A specific, simple and sensitive UHPLC-ESI-MS/MS method was developed and validated for the pharmacokinetic study of corynoline
Hom-Lay Wang et al.
Journal of periodontology, 85(12), 1693-1701 (2014-06-27)
The purpose of this multicenter randomly controlled clinical trial was to compare two acellular dermal matrix (ADM) materials produced by different processing techniques, freeze-dried (FDADM) and solvent-dehydrated (SDADM) ADM, in their ability to correct Miller Class I and II recession

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service