Skip to Content
Merck
All Photos(1)

Key Documents

PHL89616

Cyanidin 3-glucoside chloride

phyproof® Reference Substance

Synonym(s):

Kuromanin chloride, Asterin, Chrysanthemin, Cyanidin 3-O-glucoside chloride, Glucocyanidin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC3=C([O+]=C2C4=CC(O)=C(O)C=C4)C=C(O)C=C3O)O[C@H](CO)[C@H]1O.[Cl-]

InChI

1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17-,18+,19-,21-;/m1./s1

InChI key

YTMNONATNXDQJF-UBNZBFALSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, and inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

It is a chloride salt of the anthocyanidin glycoside, cyanidin-3-glucoside. Cyanidin 3-glucoside is the major anthocyanin found in legumes, such as black soybean, common bean, cowpea, and lentils. It is known to show antioxidant, anti-inflammatory, antitumor, and antihypertensive properties.

Biochem/physiol Actions

Anthocyanin. Antioxidant. Studied for its neuroprotective property.

Other Notes

This compound is commonly found in plants of the genus: echinacea sambucus zingiber echinacea sambucus zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katia Arena et al.
Analytical and bioanalytical chemistry, 415(13), 2371-2382 (2022-07-15)
In this work, a comprehensive two-dimensional liquid chromatography system, comprised of a ZIC-HILIC and C18 columns in the first and second dimensions, respectively, was tuned and employed for attaining high resolution profiles of the polyphenolic pattern in seven commercial berry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service