Skip to Content
Merck
All Photos(1)

Key Documents

63677

Supelco

(−)-Menthone

analytical standard

Synonym(s):

(1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2041368
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D −28±2°, neat

optical purity

enantiomeric ratio: ≥90:10 (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.45 (lit.)
n20/D 1.451

bp

207-210 °C (lit.)

density

0.893 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@@H]1CC[C@@H](C)CC1=O

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m1/s1

InChI key

NFLGAXVYCFJBMK-BDAKNGLRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha
Highly purified (-)-menthone. Resolving agent for diols via acetalization ; Menthone-derived spirocyclic 1,3-dioxane-4,6-diones are used in asymmetric Diels-Alder and (2+2) cycloadditions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Harada, A. Oku
Synlett, 95-95 (1994)
M. Sato et al.
Tetrahedron, 47, 7271-7271 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service