Skip to Content
Merck
All Photos(1)

Key Documents

59878

Supelco

Lithium Standard for IC

TraceCERT®, 1000 mg/L Li+ in nitric acid (nominal concentration), Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

1000 mg/L Li+ in nitric acid (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Ion Chromatography in Biomedical Research: Varao Moura A et al. conducted a study to determine ions in Caenorhabditis elegans using ion chromatography, highlighting the use of lithium standards in the precise quantification of ions for biological and pharmaceutical research. This method offers vital insights into the metabolic processes within model organisms, facilitating advancements in biochemistry and pharmacology (Varao Moura A et al., 2022).

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L Li+ in ≤0.1% nitric acid, prepared with high purity Li2CO3, HNO3 and water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Metrosep C Supp 2 column and acidic binary IC eluent accurately determine ammonium in acidic solutions, meeting pharmacopeia regulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service