Skip to Content
Merck
All Photos(1)

Key Documents

48952

Supelco

Glass Plate

Selectophore, plan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

product line

Selectophore

form

plan

size

95 mm × 200 mm

General description

Visit our Sensor Applications portal to learn more.

Application

for the preparation of ion-selective membranes, in combination with glass rings (Cat. No. 48953, 48954) and vial (Cat. No. 85795)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woo-Jae Cho et al.
Sensors (Basel, Switzerland), 19(24) (2019-12-19)
In closed hydroponics, fast and continuous measurement of individual nutrient concentrations is necessary to improve water- and nutrient-use efficiencies and crop production. Ion-selective electrodes (ISEs) could be one of the most attractive tools for hydroponic applications. However, signal drifts over

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service