Skip to Content
Merck
All Photos(2)

Key Documents

34194

Supelco

Dipentyl phthalate-3,4,5,6-d4

analytical standard

Synonym(s):

Di-n-pentyl phthalate-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D4H22O4
CAS Number:
Molecular Weight:
310.42
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCCCCC)c(c1[2H])C(=O)OCCCCC

InChI

1S/C18H26O4/c1-3-5-9-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-10-6-4-2/h7-8,11-12H,3-6,9-10,13-14H2,1-2H3/i7D,8D,11D,12D

InChI key

IPKKHRVROFYTEK-CXRURWBMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dipentyl phthalate-3,4,5,6-d4 may be used as a surrogate standard for investigating organophosphate flame retardants in food matrices using gas chromatography-triple quadrupole mass spectrometer (GC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rice ingestion is a major pathway for human exposure to organophosphate flame retardants (OPFRs) in China.
Zhang X, et al.
Journal of Hazardous Materials, 318, 686-693 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service