Skip to Content
Merck
All Photos(2)

Key Documents

32942

Supelco

Robenidine-d8 hydrochloride

VETRANAL®, analytical standard

Synonym(s):

1,3-Bis[(4-chlorobenzylidene)amino]guanidine-d8 monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D8H5Cl2N5 · HCl
CAS Number:
Molecular Weight:
378.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+8

SMILES string

Cl.[2H]c1c([2H])c(\C=N\NC(=N)N\N=C\c2c([2H])c([2H])c(Cl)c([2H])c2[2H])c([2H])c([2H])c1Cl

InChI

1S/C15H13Cl2N5.ClH/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12;/h1-10H,(H3,18,21,22);1H/b19-9+,20-10+;/i1D,2D,3D,4D,5D,6D,7D,8D;

InChI key

LTWIBTYLSRDGHP-YVXDQVGZSA-N

General description

Robenidine-d8 hydrochloride is the salt of deuterated analog of coccidiostat compound, robenidine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Robenidine-d8 hydrochloride may be used as a precursor for the preparation of robenidine-d8 which can be used as an analytical standard for the determination of robenidine in eggs and animal feed by liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of fifteen coccidiostats in feed at carry-over levels using liquid chromatography-mass spectrometry.
Pietruk K, et al.
Journal of Pharmaceutical and Biomedical Analysis, 112(1-2), 50-59 (2015)
Development and validation of a multi-residue liquid chromatography-tandem mass spectrometry confirmatory method for eleven coccidiostats in eggs.
Galarini R, et al.
Analytica Chimica Acta, 700(1-2), 167-176 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service