Skip to Content
Merck
All Photos(4)

Key Documents

2GFHSRMKB

Roche

KAPA2G Fast HotStart ReadyMix

2 ×

Synonym(s):

PCR, hotstart PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

usage

sufficient for 100 reactions
sufficient for 500 reactions

Quality Level

shelf life

≤18 mo.

feature

Fast PCR
Multiplex PCR
dNTPs included
hotstart

packaging

kit of 1.25 mL (100 x 25 μL rxn; KK5603)
kit of 6.25 mL (500 x 25 μL rxn; KK5601)

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

PCR: suitable

input

purified DNA

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA2G Fast DNA Polymerase is a second-generation (2G) enzyme engineered for higher processivity and speed, offering significantly faster extension rates than wild-type Taq polymerase. In addition to speed, KAPA2G Fast provides higher yields and sensitivity than competitor enzymes across a broad range of targets.
KAPA2G Fast DNA Polymerase Kits are designed for fast PCR, in which total reaction times are 20–70% shorter than those of conventional PCR assays performed with wild-type Taq DNA polymerase. This can be achieved without sacrificing reaction performance, and does not require specialized PCR consumables or thermocyclers.
KAPA2G Fast HotStart® DNA Polymerase is supplied in a convenient 2X ReadyMix format, containing all reaction components except primers and template.

Application

KAPA2G Fast HotStart® ReadyMix has been used for:
  • Routine PCR
  • Fast PCR
  • Reverse transcription polymerase chain reaction (RT-PCR)
  • Genotyping and fluorescent PCR

Biochem/physiol Actions

KAPA2G Fast HotStart® DNA Polymerase generated DNA fragments may be used for routine downstream analyses or applications, including restriction enzyme digestion, cloning, and sequencing. Like wild-type Taq, KAPA2G Fast has 5′→3′ polymerase and 5′→3′ exonuclease activities, but no 3′→5′ exonuclease (proofreading) activity. The fidelity of KAPA2G Fast is similar to that of wild-type Taq; it has an error rate of approximately 1 error per 1.7 x 105 nucleotides incorporated.

Features and Benefits

Reduce cycling times up to 75% :
  • Extension times as low as 1 sec per kilobase
  • Broad coverage of both AT- and GC-rich targets

High speed, higher performance :

  • Single copy sensitivity from complex targets
  • Increased yields

Quick Notes :
  • KAPA2G Fast HotStart ReadyMix PCR Kits contain the engineered KAPA2G Fast HotStart DNA Polymerase, developed for fast PCR.
  • Use 1 sec extension time for amplicons <1 kb and 15 sec/kb for longer amplicons, and save 20–70% of total reaction time.
  • No need for specialized instrumentation or PCR consumables.
  • Optimized buffer system offers improved yields, specificity and sensitivity, facilitating efficient primer annealing across a wide range of primer lengths, GC contents, and melting temperatures.
  • For high reaction efficiency, do not exceed 25 μL reaction volumes.
  • KAPA2G Fast HotStart ReadyMix contains 1.5 mM MgCl2 at 1X.
  • KAPA2G Fast HotStart ReadyMix with dye includes two inert tracking dyes to allow direct loading onto agarose gels.Reaction products are 3′-dA-tailed and may be cloned into TA cloning vectors.

Quality

Each batch of KAPA2G Fast DNA Polymerase is confirmed to contain <2% contaminating protein (Agilent Protein 230 Assay). KAPA2G Fast ReadyMix is subjected to stringent quality control tests, is free of contaminating exo- and endonuclease activity, and meets strict requirements with respect to DNA contamination levels.

Preparation Note

Always ensure that the product has been fully thawed and mixed before use. Reagents may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that all components have been handled carefully and not contaminated, the kit is not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that reagents stored at temperatures above -20°C are more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

HOTSTART is a registered trademark of Molecular BioProducts, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • KAPA2G Fast HotStart® DNA Polymerase 0.5 U per 25 µL reaction

  • MgCl2 1.5mM at 1X

  • dNTP Mix 0.2 mM of each dNTP at 1X

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An antisense RNA capable of modulating the expression of the tumour suppressor microRNA-34a.
bioRxiv, 234310-234310 (2017)
Quantification of nascent transcription by bromouridine immunocapture nuclear run-on RT-qPCR.
Roberts T C, et al.
Nature Protocols, 10(8), 1198-1198 (2015)
Khanit Sa-Ngiamsuntorn et al.
Current protocols in stem cell biology, 42, 4A-4A (2017-08-15)
This unit describes protocols to develop hepatocyte-like cells (HLCs) starting from mesenchymal stem cells (MSCs) as a natural host for hepatitis C virus (HCV). These include the preparation of MSCs from bone marrow, the reprogramming of MSCs into induced pluripotent
Alan Diot et al.
Pharmacological research, 100, 24-35 (2015-07-22)
Mitophagy is a cellular mechanism for the recycling of mitochondrial fragments. This process is able to improve mitochondrial DNA (mtDNA) quality in heteroplasmic mtDNA disease, in which mutant mtDNA co-exists with normal mtDNA. In disorders where the load of mutant
Ivana Kurelac et al.
Nature communications, 10(1), 903-903 (2019-02-24)
Converting carcinomas in benign oncocytomas has been suggested as a potential anti-cancer strategy. One of the oncocytoma hallmarks is the lack of respiratory complex I (CI). Here we use genetic ablation of this enzyme to induce indolence in two cancer

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service