Skip to Content
Merck
All Photos(4)

Key Documents

MAB1072

Sigma-Aldrich

Anti-Erythropoietin Antibody, clone 1C8

clone 1C8, Chemicon®, from mouse

Synonym(s):

erythropoietin, Epoetin, epoetin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

clone

1C8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Erythropoietin (Epo) is a glycoprotein consisting of one 165-amino acid single-chain polypeptide with two internal disulfide bonds. When fully glycosylated, the molecular weight of Epo is approximately 34 kDa. Normally produced by the liver and kidney, Epo is the principle regulator of red blood cell production by stimulating the division and differentiation of bone marrow erythroid progenitor cells in response to anemia and hypoxia.

Specificity

Reacts with human Erythropoietin (Epo).

Immunogen

Recombinant human Epo.

Application

Detect Erythropoietin using this Anti-Erythropoietin Antibody, clone 1C8 validated for use in ELISA, IF & WB.
ELISA:
For the determination of Epo concentration in the range of 2-200 ng/mL. Use at 5 ug/mL to coat plate.

Immunosorbent preparation

Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Metabolic Hormones & Receptors

Quality

Evaluated by Western Blot on Mouse kidney lysates.

Western Blot Analysis:
1:1000 dilution of this antibody detected glycosylated EPO on 10 µg of Mouse kidney lysates.

Target description

34 kDa (glycosylated)

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2b in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Produced by kidney or liver of adult mammals and by liver of fetal or neonatal mammals.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas H Sanderson et al.
PloS one, 8(11), e78627-e78627 (2013-11-14)
Recent advancements in isolation techniques for cytochrome c (Cytc) have allowed us to discover post-translational modifications of this protein. We previously identified two distinct tyrosine phosphorylated residues on Cytc in mammalian liver and heart that alter its electron transfer kinetics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service