Skip to Content
Merck
All Photos(1)

Key Documents

8.20572

SAFC

Fluorene

for synthesis

Synonym(s):

Fluorene, Diphenylenemethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352005
EC Index Number:
201-695-5

vapor pressure

13 hPa ( 146 °C)

Quality Level

200
300

Assay

≥95.0% (GC)

form

powder

mp

112-116 °C (External MSDS)

transition temp

flash point 151 °C (External MSDS)

storage temp.

2-30°C

InChI

1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2

InChI key

NIHNNTQXNPWCJQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Melting range (lower value): ≥ 113 °C
Melting range (upper value): ≤ 115 °C
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service