Skip to Content
Merck
All Photos(1)

Key Documents

8.18194

Sigma-Aldrich

Sodium methylate

(30% solution in methanol) for synthesis

Synonym(s):

Sodium methoxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107
NACRES:
NA.22

vapor pressure

34 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

420 °C (Methanol)

expl. lim.

5.5-44 % (v/v) Methanol)

pH

11 (20 °C, 20 g/L in H2O)

bp

92 °C/1013 hPa

mp

2-6 °C

transition temp

flash point 33 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • Approach to synthesis and study of pharmacological effects of new 1, 3, 5-triazine derivatives: This research includes the use of sodium methylate in methanol medium for synthesizing new triazine derivatives, predicting their pharmacological effects using GUSAR software (Levshukova et al., 2024).

  • Exothermic barrier-free carboxylation of alkali methylates: Investigates the interaction of sodium methylate with CO2 to synthesize dialkyl carbonates, presenting a high-yield method rationalized through molecular dynamics (Chaban, 2023).

Analysis Note

Assay (acidimetric): 28.5 - 31.5 %
Density (d 20 °C/ 4 °C): 0.968 - 0.972

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service