Skip to Content
Merck
All Photos(1)

Key Documents

8.14575

Sigma-Aldrich

Citronellal

for synthesis

Synonym(s):

Citronellal, 3.7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352114
EC Index Number:
203-376-6
NACRES:
NA.22

vapor pressure

14 hPa ( 88 °C)

Quality Level

Assay

≥93.5% (GC)

form

liquid

autoignition temp.

202 °C

potency

2420 mg/kg LD50, oral (Rat)
>2500 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-4.5 % (v/v)

pH

7 ( in H2O)

bp

208 °C/1013 hPa

transition temp

flash point 77 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3/t10-/m1/s1

InChI key

NEHNMFOYXAPHSD-SNVBAGLBSA-N

Analysis Note

Assay (GC, area%): ≥ 93.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.851 - 0.854
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service