Skip to Content
Merck
All Photos(1)

Key Documents

553031

Sigma-Aldrich

RAGE Antagonist Peptide, RAP

The RAGE Antagonist Peptide, RAP controls the biological activity of RAGE.

Synonym(s):

RAGE Antagonist Peptide, RAP, ELKVLMEKEL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H101N13O17S
Molecular Weight:
1272.55
UNSPSC Code:
12352111
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

potency

2 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 5 mg/mL

shipped in

dry ice

storage temp.

−20°C

General description

An S100P-derived peptide based competitive antagonist for receptor for advanced glycation end product (RAGE). Shown to disrupt the interaction between RAGE and its ligands, such as S100P, S100A4 and HMGB-1 in binding assays and in multiple cancer cell lines (~2 µM). Also, blocks RAGE-dependent NF-kB activation in MPanc96, MOH and HPAF II tumor cell lines. Systemic administration of RAP also diminishes NF-kB signaling in vivo and significantly reduces glioma tumor growth in murine models.

Biochem/physiol Actions

Primary Target
RAGE
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-ELKVLMEKEL-NH2

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Arumugam, T., et al. 2012. Clin Cancer Res.18, 4356.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Choltus et al.
International journal of molecular sciences, 22(15) (2021-08-08)
Maternal smoking is a risk factor of preterm prelabor rupture of the fetal membranes (pPROM), which is responsible for 30% of preterm births worldwide. Cigarettes induce oxidative stress and inflammation, mechanisms both implicated in fetal membranes (FM) weakening. We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service