Skip to Content
Merck
All Photos(1)

Key Documents

1.03880

Millipore

Yersinia Selective Supplement (CIN)

selective for bacteria: Yersinia Y. enterocolitica, pkg of 10 vials, for use with GranuCult® prime CIN (Cefsulodin, Irgasan, Novobiocin) Agar

Synonym(s):

Yersinia Selective Supplement, Yersinia Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

form

liquid (vial)

Quality Level

composition

Cefsulodin, 7.5 mg per vial
Irgasan, 2 mg per vial
Novobiocin, 1.25 mg per vial

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

compatibility

for use with GranuCult® prime CIN (Cefsulodin, Irgasan, Novobiocin) Agar

storage temp.

2-8°C

suitability

selective for bacteria: Yersinia Y. enterocolitica

General description

Yersinia Selective Supplement (CIN) consists of a lyophilized mixture of three inhibitors (Cefsulodin, Irgasan, and Novobiocin). The supplement inhibits the growth of accompanying flora encountered in the cultivation of Yersinia, particularly Y. enterocolitica.

Application

Yersinia Selective Supplement is an additive for the preparation of Yersinia selective agar for the enrichment of Yersinia spp.

Compatibility

For use with GranuCult® prime CIN (Cefsulodin, Irgasan, Novobiocin) Agar (1.03871)

Analysis Note

Yersinia Selective Supplement (CIN) should be used with liquid Yersinia selective agar base at a temperature of 45-50 degrees.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service