Skip to Content
Merck
All Photos(1)

Key Documents

1.03859

Millipore

GranuCult® prime KLIGLER Agar

acc. to FDA-BAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Quality Level

form

powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

nonselective for

General description

GranuCult® prime KLIGLER Agar is a nutrient rich medium which is designed for the identification of Gram-negative bacteria, mainly Enterobacteriaceae, on the base of their ability to ferment glucose and lactose (carbohydrate fermentation) and to release sulfides (H2S production). The rich carbon and carbohydrate sources like peptone, meat extract and glucose enable rapid growth of microorganisms present in the test sample.

Application

GranuCult® prime KLIGLER Agar is used to identify gram-negative bacteria from the test samples.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service