Skip to Content
Merck
All Photos(1)

Key Documents

Z208213

Sigma-Aldrich

Rubbermaid® heavy-duty lab carts

Model B, H × W × L 33 1/4 in. × 25 7/8 in. × 45 1/4 in., black, cart total capacity 500 lb. , 250 lb. per shelf

Synonym(s):

heavy-duty cart, rubbermaid cart, rubbermaid carts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
JA.01

manufacturer/tradename

Model B

H × W × L

33 1/4 in. × 25 7/8 in. × 45 1/4 in.

cart total capacity

500 lb. , 250 lb. per shelf

color

black

Looking for similar products? Visit Product Comparison Guide

General description

Duramold® construction holds up under rugged use. Seamless construction, no sharp edges. Easy to clean. Maneuverable. Optional middle shelf sold separately for models B and C.

Legal Information

Duramold is a registered trademark of Rubbermaid Commercial Products, Inc.
Rubbermaid is a registered trademark of Rubbermaid Commercial Products, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service