Skip to Content
Merck
All Photos(1)

Key Documents

W410100

Sigma-Aldrich

Mango furanone

natural, ≥98%, FG

Synonym(s):

2,5 Dimethyl furan-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
FEMA Number:
4101
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.119
NACRES:
NA.21

biological source

botanical

Quality Level

grade

FG
Halal
Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; fruity

InChI

1S/C6H8O2/c1-4-3-6(7)5(2)8-4/h3,5H,1-2H3

InChI key

ASOSVCXGWPDUGN-UHFFFAOYSA-N

Legal Information

For R&D use and/or Food Manufacture Only.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.0 °F

Flash Point(C)

51.67 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service