Skip to Content
Merck
All Photos(1)

Key Documents

W345001

Sigma-Aldrich

2,5-Dimethyl-2,5-dihydroxy-1,4-dithiane

95%, FG

Synonym(s):

Meaty dithiane, 1-Mercaptopropanone (dimer), 2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane, 2,5-Dimethyl-1,4-dithiane-2,5-diol, 2,5-Dimethyl-2,5-dihydroxy-p-dithiane, Cyclodithalfarol-705

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2S2
CAS Number:
Molecular Weight:
180.29
FEMA Number:
3450
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
15.006
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

95%

mp

124-127 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

CC1(O)CSC(C)(O)CS1

InChI

1S/C6H12O2S2/c1-5(7)3-10-6(2,8)4-9-5/h7-8H,3-4H2,1-2H3

InChI key

NHKIYYMFGJBOTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service