Skip to Content
Merck
All Photos(1)

Key Documents

P10402

Sigma-Aldrich

Perfluoropropyl iodide

98%

Synonym(s):

1-Iodoperfluoropropane, Heptafluoro-1-iodopropane, Heptafluoropropyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2I
CAS Number:
Molecular Weight:
295.93
Beilstein:
1758822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7.08 psi ( 20 °C)

Assay

98%

contains

copper as stabilizer

refractive index

n20/D 1.328 (lit.)

bp

41 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C3F7I/c4-1(5,2(6,7)8)3(9,10)11

InChI key

XTGYEAXBNRVNQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Dodd et al.
Drug and chemical toxicology, 21(2), 137-149 (1998-05-23)
Trifluoroiodomethane (CF3I) and 1,1,2,2,3,3,3-heptafluoro-1-iodopropane (C3F7I) have been considered as replacement candidates for halon fire suppressants due to their excellent fire extinguishant capabilities and low ozone depletion potential compared to halon fire extinguishants in use currently. As part of the process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service