Skip to Content
Merck
All Photos(1)

Key Documents

BAF500000100

SP Bel-Art® Techni-Dome 360 Fume Hood

polycarbonate, W × D 60 cm (22 in.) × 60 cm (22 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

material

clear
polycarbonate

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F50000-0100

W × D

60 cm (22 in.) × 60 cm (22 in.)

capacity

0.5 mL
65 L

General description

The Techni-Dome® Fume Hood has a 50cm (20") ID and 45cm (18") height, making it ideal for simple micropipetting processes, blending of chemicals, handling of small biohazard items or interactive museum/educational displays.
  • Compact; fits on crowded benchtops
  • Clear, unrestricted viewing from all angles
  • Polycarbonate plastic; resistant to a wide range of chemicals
  • Smooth interior for easy cleaning
  • High impact strength and scratch-resistance
  • Built-in handles makes it portable and suited for field use
  • Both halves of the unit nest for compact storage
  • Stepped outlet connects to standard exhaust systems with 4 or 6" vent piping
  • Overall volume, 65 liters (4000 cu. in.)
  • Overall dimensions 60W x 60cm H (22"W x 22"H)

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Techni-Dome is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service