Skip to Content
Merck
All Photos(1)

Documents

908673

Sigma-Aldrich

Nile blue acrylamide

Synonym(s):

Fluoresent monomer, Nile blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22N3O2Cl
CAS Number:
Molecular Weight:
407.89
MDL number:
UNSPSC Code:
41105319
NACRES:
NA.23

form

crystals

color

dark blue to purple

storage temp.

−20°C

Application

Nile blue acrylamide is a fluorescent monomer for synthesizing fluorescent polymers. Fluorescent polymers are especially useful in the preparation of fluorescent microspheres or nanoparticles, for bioimaging or drug delivery research.

Storage and Stability

Protect from light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Tenuta et al.
PloS one, 6(10), e25556-e25556 (2011-10-15)
Cells act as extremely efficient filters for elution of unbound fluorescent tags or impurities associated with nanoparticles, including those that cannot be removed by extensive cleaning. This has consequences for quantification of nanoparticle uptake and sub-cellular localization in vitro and
Fluorescent and chemico-fluorescent responsive polymers from dithiomaleimide and dibromomaleimide functional monomers.
Robin M P et al.
Chemical Science, 5, 2717-2723 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service