Skip to Content
Merck
All Photos(1)

Key Documents

751316

Sigma-Aldrich

3-epi-25-Hydroxyvitamin D3 (6,19,19-d3)

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D3H41O2
Molecular Weight:
403.66
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

off-white to beige

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1CC[C@@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23-,24-,25+,27-/m1/s1/i1D2,12D

InChI key

JWUBBDSIWDLEOM-BOFGWHLFSA-N

General description

3-epi-25-Hydroxyvitamin D3 (C3-epimer) is a metabolite of vitamin D3. It is an epimer of 25-hydroxyvitamin D3 which differs only at the C-3 position wherein -OH group at C-3 carbon is in the α orientation rather than in the β orientation.
3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.

Application

3-epi-25-Hydroxyvitamin D3 can be used as an analytical standard for the quantitative analysis of 3-epi-25-hydroxyvitamin D3 content in biological fluids via LC-MS/MS.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overestimation of 25-hydroxyvitamin D3 by increased ionisation efficiency of 3-epi-25-hydroxyvitamin D3 in LC-MS/MS methods not separating both metabolites as determined by an LC-MS/MS method for separate quantification of 25-hydroxyvitamin D3, 3-epi-25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 in human serum
van den Ouweland JMW, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 967, 195-202 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service