Skip to Content
Merck
All Photos(2)

Key Documents

720410

Sigma-Aldrich

3,5-Dimethylisoxazol-4-yl-4-boronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8BNO3
CAS Number:
Molecular Weight:
140.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

mp

82-86 °C

storage temp.

2-8°C

SMILES string

Cc1noc(C)c1B(O)O

InChI

1S/C5H8BNO3/c1-3-5(6(8)9)4(2)10-7-3/h8-9H,1-2H3

InChI key

DIIFZCPZIRQDIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service