Skip to Content
Merck
All Photos(1)

Key Documents

53588

Sigma-Aldrich

Homopiperazine-1,4-bis(2-ethanesulfonic acid)

Synonym(s):

Hexahydro-1H-1,4-diazepine-1,4-bis(2-ethanesulfonic acid), Homo-PIPES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20N2O6S2
CAS Number:
Molecular Weight:
316.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

impurities

≤5% water

SMILES string

OS(=O)(=O)CCN1CCCN(CC1)CCS(O)(=O)=O

InChI

1S/C9H20N2O6S2/c12-18(13,14)8-6-10-2-1-3-11(5-4-10)7-9-19(15,16)17/h1-9H2,(H,12,13,14)(H,15,16,17)

InChI key

QZTMPPUJIVQNKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Homopiperazine-1,4-bis(2-ethanesulfonic acid) is used as a buffer in experiments involving plant cells at low pH.

Application

  • Evaluation of buffers toxicity in plant cells: Homopiperazine-1,4-bis(2-ethanesulfonic acid) was evaluated for its suitability as a buffer in tobacco cell studies at low pH, highlighting its low toxicity and effectiveness in maintaining stable pH environments crucial for plant cell biology experiments (Borgo, 2017).
  • Proteomic analysis under stress conditions: This buffer was used in maintaining pH during the proteomic analysis of radicles from seeds treated with aluminum, showcasing its utility in complex biological assays and environmental stress studies (Okekeogbu et al., 2014).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of buffers toxicity in tobacco cells: Homopiperazine-1, 4-bis (2-ethanesulfonic acid) is a suitable buffer for plant cells studies at low pH
Borgo L
Plant Physiology and Biochemistry, 115, 119-125 (2017)
Doris Pester et al.
PloS one, 7(3), e32583-e32583 (2012-03-14)
Pathogen entry through host blossoms is the predominant infection pathway of the gram-negative bacterium Erwinia amylovora leading to manifestation of the disease fire blight. Like in other economically important plant pathogens, E. amylovora pathogenicity depends on a type III secretion
Florian Schmidt et al.
Nature, 562(7727), 380-385 (2018-10-05)
The ability to record transcriptional events within a cell over time would help to elucidate how molecular events give rise to complex cellular behaviours and states. However, current molecular recording technologies capture only a small set of defined stimuli. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service