Skip to Content
Merck
All Photos(1)

Key Documents

489573

Sigma-Aldrich

Guanidine-d5 deuteriochloride

98 atom % D

Synonym(s):

Guanidine-d5 mono(hydrochloride-d)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2NC(=ND)ND2
CAS Number:
Molecular Weight:
101.57
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mp

185-189 °C (lit.)

mass shift

M+6

SMILES string

Cl[2H].[2H]\N=C(/N([2H])[2H])N([2H])[2H]

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H/i/hD6

InChI key

PJJJBBJSCAKJQF-YIKVAAQNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Tsirigotaki et al.
Scientific reports, 7(1), 2456-2456 (2017-05-28)
Prolyl oligopeptidase (PREP) is conserved in many organisms across life. It is involved in numerous processes including brain function and neuropathology, that require more than its strict proteolytic role. It consists of a seven-bladed β-propeller juxtaposed to a catalytic α/β-hydrolase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service