Skip to Content
Merck
All Photos(1)

Key Documents

468053

Sigma-Aldrich

L-(−)-chiro-Inositol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]20/D −60°, c = 1.3 in H2O

mp

230 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4-,5+,6+/m1/s1

InChI key

CDAISMWEOUEBRE-SHFUYGGZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuepeng Wang et al.
The Analyst, 144(8), 2611-2617 (2019-03-06)
The discrimination of chirality by field effect transistors (FETs) is a great challenge because enantiomers possess exactly identical charge and hence chiral recognition is solely related to spatial effects, while the working principle of most FET sensors is based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service