Skip to Content
Merck
All Photos(1)

Key Documents

408549

Sigma-Aldrich

Z-His-OH

99%

Synonym(s):

Nα-Carbobenzyloxy-L-histidine, Nα-Z-L-Histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3O4
CAS Number:
Molecular Weight:
289.29
Beilstein:
92342
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −24°, c = 1% in 6 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

168 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](Cc1c[nH]cn1)NC(=O)OCc2ccccc2

InChI

1S/C14H15N3O4/c18-13(19)12(6-11-7-15-9-16-11)17-14(20)21-8-10-4-2-1-3-5-10/h1-5,7,9,12H,6,8H2,(H,15,16)(H,17,20)(H,18,19)/t12-/m0/s1

InChI key

WCOJOHPAKJFUDF-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service