Skip to Content
Merck
All Photos(1)

Key Documents

326267

Sigma-Aldrich

2,3,4,5-Tetrafluorobenzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6F4CO2H
CAS Number:
Molecular Weight:
194.08
Beilstein:
2052670
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

85-87 °C (lit.)

SMILES string

OC(=O)c1cc(F)c(F)c(F)c1F

InChI

1S/C7H2F4O2/c8-3-1-2(7(12)13)4(9)6(11)5(3)10/h1H,(H,12,13)

InChI key

SFKRXQKJTIYUAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4,5-Tetrafluorobenzoic acid was used in preparation of series of new organotin(IV) 2,3,4,5-tetrafluorobenzoic acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Self-assembled diorganotin (IV) moieties with 2, 3, 4, 5-tetrafluorobenzoic acid: Syntheses, characterizations and crystal structures.
Ma C, et al.
Inorgorganica Chimica Acta, 359(13), 4179-4190 (2006)
Jihye Kim et al.
Analytical and bioanalytical chemistry, 411(5), 1067-1073 (2018-12-20)
Single-drop microextraction (SDME) and large-volume sample stacking using an electroosmotic flow pump (LVSEP) were coupled with capillary electrophoresis/mass spectrometry (CE/MS) for sample cleanup and preconcentration. Without filtration or centrifugation of a soil sample containing debris, SDME using a pentanol acceptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service