Skip to Content
Merck
All Photos(1)

Key Documents

158224

Sigma-Aldrich

N-(4-Methoxybenzylidene)-4-butylaniline

98%

Synonym(s):

MBBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4N=CHC6H4OCH3
CAS Number:
Molecular Weight:
267.37
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid crystal

refractive index

n20/D 1.5496 (lit.)

density

1.027 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccc(cc1)\N=C\c2ccc(OC)cc2

InChI

1S/C18H21NO/c1-3-4-5-15-6-10-17(11-7-15)19-14-16-8-12-18(20-2)13-9-16/h6-14H,3-5H2,1-2H3/b19-14+

InChI key

FEIWNULTQYHCDN-XMHGGMMESA-N

General description

N-(4-Methoxybenzylidene)-4-butylaniline (MBBA) is a nematic liquid crystal molecule that can be prepared by condensing distilled n-butylaniline and methoxybenzaldehyde. It can be used as an anisotropic solvent which has an isotropic transition temperature of 315k. It is majorly used in the development of liquid crystal cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukas Jackalin et al.
Physical chemistry chemical physics : PCCP, 20(34), 22187-22196 (2018-08-18)
Direct dipolar spin couplings are informative and sensitive probes for a wide range of dynamic processes and structural properties at atomic, molecular and supramolecular levels in liquid crystals and other anisotropic materials. Usually, heteronuclear 13C-1H dipolar couplings in liquid crystals
Absorption anisotropy and molecular association of some ionic dyes in liquid crystalline solution
Ghanadzadeh A and Zakerhamidi MS
Journal of Molecular Liquids, 109(3), 149-154 (2004)
Luis A Tercero Espinoza et al.
Langmuir : the ACS journal of surfaces and colloids, 20(6), 2375-2385 (2005-04-20)
We report the orientational behavior of nematic phases of 4-cyano-4'-pentylbiphenyl (5CB) on cationic, anionic, and nonionic surfaces before and after contact of these surfaces with solutions containing the negatively charged vesicular stomatitis virus (VSV). The surfaces were prepared on evaporated
Karel Kouřil et al.
Physical chemistry chemical physics : PCCP, 19(19), 11793-11801 (2017-04-27)
We report the NMR of the molecular endofullerenes H
Flexoelectro-optic effect in a hybrid nematic liquid crystal cell
Ponti S, et al.
Liq. Cryst., 26(8), 1171-1177 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service