Skip to Content
Merck
All Photos(2)

Key Documents

126527

Sigma-Aldrich

2,6-Dimethylmorpholine

97%

Synonym(s):

2,6-Dimethylmorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
103036
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.446 (lit.)

bp

147 °C (lit.)

mp

−85 °C (lit.)

density

0.935 g/mL at 25 °C (lit.)

SMILES string

CC1CNCC(C)O1

InChI

1S/C6H13NO/c1-5-3-7-4-6(2)8-5/h5-7H,3-4H2,1-2H3

InChI key

HNVIQLPOGUDBSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-2,6-Dimethylmorpholine (DMMP) has been used in the preparation of microporous material composed of hybrid T2 and T3 supertetrahedral clusters.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Mirvish et al.
Cancer research, 43(6), 2550-2554 (1983-06-01)
We reported previously that mice exposed to atmospheric NO2 contained a nitrosating agent (NSA) that reacted with morpholine in aqueous methanol homogenates of the mice to give N-nitrosomorpholine. We have now found that N-nitrosomorpholine was also produced by reacting morpholine
Xiaohui Han et al.
Chemical communications (Cambridge, England), 50(7), 796-798 (2013-12-03)
Solvothermal reactions of S, Ga and Sn using cis-2,6-dimethylmorpholine (DMMP) as an organic template, in the presence of a trace amount of H2O at 180 °C, generated microporous [C6H14NO]14.4[Ga6.40Sn21.60S52O8] material composed of hybrid T2 and T3 supertetrahedral clusters with a
Natalia Szkaradek et al.
Anti-cancer agents in medicinal chemistry, 19(16), 1949-1965 (2019-04-06)
Natural plant metabolites and their semisynthetic derivatives have been used for years in cancer therapy. Xanthones are oxygenated heterocyclic compounds produced as secondary metabolites by higher plants, fungi or lichens. Xanthone core may serve as a template in the synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service