Skip to Content
Merck
All Photos(1)

Key Documents

112437

Sigma-Aldrich

4-Ethylpyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
106481
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.501 (lit.)

bp

168 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

CCc1ccncc1

InChI

1S/C7H9N/c1-2-7-3-5-8-6-4-7/h3-6H,2H2,1H3

InChI key

VJXRKZJMGVSXPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethylpyridine undergoes metal-ligand interaction with Zn-porphyrin which can be studied by mass spectrometry.

Application

4-Ethylpyridine is used to study the influence of counter-anion concentration on analyte retention.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R LoBrutto et al.
Journal of chromatography. A, 913(1-2), 189-196 (2001-05-18)
The influence of acid and salt concentration in the mobile phase on the retention of basic analytes has been studied. An increase in the retention of fully protonated analytes with increasing the concentration of inorganic additives was found. The addition
Monica Brivio et al.
Lab on a chip, 5(10), 1111-1122 (2005-09-22)
Two simple interfaces were designed and realized, enabling on-line coupling of microfluidics reactor chips to a nanoflow electrospray ionization (NESI) time-of-flight (TOF) mass spectrometer (MS). The interfaces are based on two different approaches: a monolithically integrated design, in which ionization
Slavko Klobčar et al.
Biomedical chromatography : BMC, 29(12), 1901-1906 (2015-05-27)
The isolation of four oxidative degradation products of atorvastatin using preparative high-performance liquid chromatography applying at least two chromatographic steps is known from the literature. In this paper it is shown that the same four impurities could be isolated from
Michael Wleklinski et al.
Physical chemistry chemical physics : PCCP, 17(28), 18364-18373 (2015-06-25)
Electrospray ionization of metal salt solutions followed by ambient heating transforms the resulting salt clusters into new species, primarily naked ionic metal clusters. The experiment is done by passing the clusters through a heated coiled loop outside the mass spectrometer
Alexandre Grand-Guillaume Perrenoud et al.
Journal of chromatography. A, 1360, 275-287 (2014-08-19)
Superficially porous particles (SPP), or core shell particles, which consist of a non-porous silica core surrounded by a thin shell of porous silica, have gained popularity as a solid support for chromatography over the last decade. In the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service