Dot/Slot (Filtration) Blotting
Dot blotting is an ideal technique for quickly assessing the levels of a target antigen across many samples at once. Also, it is a popular method for epitope mapping and screening antibodies for target specificity.
Dot/Slot (Filtration) blotting topics: possible causes and remedies:
Figure 1.Detection of transferrin on a dot blot of human serum using Immobilon® Western Chemiluminescent HRP Substrate.
Slow or No Filtration of the Sample Through the Membrane | |
---|---|
Possible Cause | Remedy |
Inadequate vacuum |
|
Membrane pores clogged |
|
Little or No Protein Observed on the Blot | |
---|---|
Possible Cause | Remedy |
Not enough protein applied to the membrane | Minimize sample dilution and filter more sample through the membrane. |
Detergents (e.g., SDS) may inhibit lower molecular weight proteins from binding to the membrane | Eliminate detergents if possible. |
Stain not sensitive enough | Use a more sensitive stain. |
Stained Blot Is Not Uniform | |
---|---|
Possible Cause | Remedy |
Membrane structure was compressed by filter paper | Place a second membrane in the blotting unit to protect the membrane receiving the samples. |
Air bubbles trapped in the interior of the membrane | Pre-wet membrane by laying it on the surface of the methanol. Immersing the membrane can entrap air. |
Membrane not pre-wet in methanol | Membrane must be pre-wet with methanol; entire membrane should change uniformly from opaque to semi-transparent. |
Air bubbles in the sample. | Carefully pipette samples into well to avoid the formation of air bubbles. |
Not enough sample volume loaded | Sample must cover the entire exposed membrane area. |
Protein Smeared Across the Top of the Membrane | |
---|---|
Possible Cause | Remedy |
Sample leaked across the wells | Make sure the blotting unit is properly assembled, closed and sealed prior to filtration. |
Protein Smeared Across the Back of the Membrane | |
---|---|
Possible Cause | Remedy |
Membrane capacity was exceeded | Reduce the amount of protein loaded into the well. |
Network error: Failed to fetch