Merck
HomeProtein PurificationSeparation Options with Sephadex®
Separation Options with Sephadex®
For group separations the medium should be selected so that the high molecular weight molecules are eluted at the void volume with minimum peak broadening or dilution and minimum time on the column. The lowest molecular weight substances should appear by the time one column volume of buffer has passed through the column. Sephadex® media are available in prepacked columns (also spin columns), microplates, and in bulk media packs (Table 5.2).
Chromatography Medium
Columns and 96-well platesLoaded
volume (mL)
Eluted
volume (mL)
Dilution
factor
Operation
Sephadex® G-25 Medium
PD SpinTrap™ G-250.07 to 0.130.07 to 0.13*No dilutionCentrifuge
PD MultiTrap™ G-250.07 to 0.130.07 to 0.13*No dilution
Centrifuge
PD MiniTrap™ G-250.2 to 0.5
0.1 to 0.5
0.1 to 0.5
1.0
No dilution
2 to 10
Centrifuge
Gravity flow
PD MidiTrap™ G-250.75 to 1.0
0.5 to 1.0
0.5 to 1.0
1.5
No dilution
1.5 to 3
Centrifuge
Gravity flow
PD-10 Desalting columns1.75 to 2.5
1.0 to 2.5
1.0 to 2.5
3.5
No dilution
1.5 to 3.5
Centrifuge
Gravity flow
Sephadex® G-10 Medium
PD MiniTrap™ G-100.1 to 0.30.51.7 to 5Gravity flow
PD MidiTrap™ G-100.4 to 1.01.21.2 to 3.0Gravity flow
Sephadex® G-25 Superfine
HiTrap® Desalting0.25
0.5
1.0
1.5 (max.)
1.0
1.5
2.0
2.0
4 (approx.)
3 (approx.)
2 (approx.)
1.3 (approx.)
Syringe/pump/system
Syringe/pump/system
Syringe/pump/system
Syringe/pump/system
2× HiTrap® Desalting3.0 (max.)4.0 to 5.01.3 to 1.7Syringe/pump/system
3× HiTrap® Desalting4.5 (max.)6.0 to 7.01.3 to 1.7Syringe/pump/system
Sephadex® G-25 Fine
HiPrep™ 26/10 Desalting
10
15 (max.)
10 to 15
15 to 20
1.0 to 1.5
1.0 to 1.3
Pump/system
Pump/system
2× HiPrep™ 26/10 Desalting30 (max.)30 to 401.0 to 1.3Pump/system
3× HiPrep™ 26/10 Desalting45 (max.)45 to 551.0 to 1.2Pump/system
4× HiPrep™ 26/10 Desalting60 (max.)60 to 701.0 to 1.2Pump/system
Table 5.2.Group Separation options with Sephadex® media
* Applied volume = eluted volume; For sample volumes less than 100 µL it is recommended to apply a stacker volume of 30 µL equilibration buffer after the sample has fully absorbed.
For convenience and reliable performance, use prepacked Sephadex columns such as HiTrap® Desalting 5 ml and HiPrep™ 26/10 Desalting.
Always use disposable columns if there is a risk of biological or radioactive contamination or when any possibility of carryover between samples is unacceptable.
Materials
Nous sommes désolés, une erreur inattendue s'est produite
Network error: Failed to fetch