Merck
Transfer of Dyes and Drugs into Cells Using EGFR-Targeted Nanosyringes.
ACS chemical neuroscience (2017-07-29)
Lizanne G Nilewski, Melissa Singh, David S Baskin, James M Tour, Martyn A Sharpe
RÉSUMÉ
Selective targeting of drug loaded nanovectors to specific epitopes highly expressed on the surface of cancer cells is a goal for nanotechnologists. We have modified our previously described PEGylated-hydrophilic carbon clusters (PEG-HCCs) so that the epidermal growth factor receptor (EGFR) binding peptide, GE11, is attached using click chemistry at the end of each PEG. The resulting nanosyringe, Pep
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Avidin–FITC from egg white, lyophilized powder