Merck
Base excision repair imbalance in colorectal cancer has prognostic value and modulates response to chemotherapy.
Oncotarget (2017-09-15)
Natalia M Leguisamo, Helena C Gloria, Antonio N Kalil, Talita V Martins, Daniel B Azambuja, Lisiane B Meira, Jenifer Saffi
RÉSUMÉ
Colorectal cancer (CRC) is prevalent worldwide, and treatment often involves surgery and genotoxic chemotherapy. DNA repair mechanisms, such as base excision repair (BER) and mismatch repair (MMR), may not only influence tumour characteristics and prognosis but also dictate chemotherapy response. Defective MMR contributes to chemoresistance in colorectal cancer. Moreover, BER affects cellular survival by repairing genotoxic base damage in a process that itself can disrupt metabolism. In this study, we characterized BER and MMR gene expression in colorectal tumours and the association between this repair profile with patients' clinical and pathological features. In addition, we exploited the possible mechanisms underlying the association between altered DNA repair, metabolism and response to chemotherapy. Seventy pairs of sporadic colorectal tumour samples and adjacent non-tumour mucosal specimens were assessed for BER and MMR gene and protein expression and their association with pathological and clinical features. MMR-deficient colon cancer cells (HCT116) transiently overexpressing
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-MPG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution