- Isolation and cultivation of Lyme disease spirochetes.
Isolation and cultivation of Lyme disease spirochetes.
The Yale journal of biology and medicine (1984-07-01)
A G Barbour
PMID6393604
RÉSUMÉ
The successful isolation and cultivation of Lyme disease spirochetes traces its lineage to early attempts at cultivating relapsing fever borreliae. Observations on the growth of Lyme disease spirochetes under different in vitro conditions may yield important clues to both the metabolic characteristics of these newly discovered organisms and the pathogenesis of Lyme disease.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
BSK-H Medium, With sodium bicarbonate, suitable for Borrelia burgdorferi (Qualified)